Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTT2 Antibody (AT10H4), PerCP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250577PCP
Description
GSTT2 Monoclonal specifically detects GSTT2 in Human samples. It is validated for Flow Cytometry.Specifications
GSTT2 | |
Monoclonal | |
PerCP | |
PBS with 0.05% Sodium Azide | |
GSTT2 | |
Recombinant human GSTT2 (1-244aa) purified from E. coli | |
100 μL | |
Cancer | |
2953 | |
Store at 4C in the dark. | |
IgG2b κ |
Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
AT10H4 | |
Flow Cytometry | |
EC 2.5.1.18, glutathione S-transferase theta 2, glutathione S-transferase theta-2, Glutathione S-transferase theta-2B, GST class-theta-2, GSTT2B, MGC182032 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction