Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTT2 Antibody (OTI3B6), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270861PE
Description
GSTT2 Monoclonal specifically detects GSTT2 in Human samples. It is validated for Flow Cytometry.Specifications
GSTT2 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
GSTT2 | |
Full length recombinant protein of human GSTT2 (NP_000845) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human | |
IgG2b |
Flow Cytometry | |
OTI3B6 | |
Flow Cytometry | |
EC 2.5.1.18, glutathione S-transferase theta 2, glutathione S-transferase theta-2, GST class-theta-2, MGC182032 | |
Mouse | |
Affinity Purified | |
Cancer | |
2953.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction