Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTT2 Monoclonal Antibody (OTI6C4), TrueMAB™, OriGene
SDP

Catalog No. 502541942
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-1942 30 μL
50-167-2991 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-1942 Supplier OriGene Supplier No. TA501883S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

GSTT2 Monoclonal antibody specifically detects GSTT2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen GSTT2
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6C4
Concentration 0.62 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Gstt2
Gene Accession No. P0CG29
Gene Alias AI266894; Glutathione S-transferase 12; glutathione S-transferase theta 2 (gene/pseudogene); glutathione S-transferase theta-2; Glutathione S-transferase Yrs-Yrs; glutathione S-transferase, theta 2; GST 12-12; GST class-theta-2; GSTT2; mGSTT2; Yrs
Host Species Mouse
Immunogen Full length human recombinant protein of human GSTT2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2953
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.