Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTT2B Antibody, Biotin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306612B
This item is not returnable.
View return policy
Description
GSTT2B Polyclonal antibody specifically detects GSTT2B in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
GSTT2B | |
Polyclonal | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
653689 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunofluorescence | |
Biotin | |
glutathione S-transferase theta 2, Glutathione S-Transferase Theta 2B (Gene/Pseudogene), Glutathione S-transferase theta-2B, GST class-theta-2, GSTT2P | |
Produced in rabbits immunized with purified, recombinant Human GSTT2B (NP_000845.1; Met1-Pro244). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction