Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GSTZ1 Antibody (044), Janelia Fluor™ 646, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290273JF646
Description
GSTZ1 Monoclonal antibody specifically detects GSTZ1 in Human samples. It is validated for ELISASpecifications
GSTZ1 | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
044 | |
ELISA | |
EC 2.5.1.18, EC 5.2.1.2, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, Glutathione S-transferase zeta 1, glutathione transferase zeta 1, GSTZ1-1maleylacetoacetate isomerase, MAAIglutathione S-aryltransferase, MAI, maleylacetone isomerase, MGC2029, S-(hydroxyalkyl)glutathione lyase | |
Rabbit immunized with purified, recombinant Human GSTZ1 (Accession#: NP_665877.1, Met1-Ala216) | |
0.1 mL | |
metabolism | |
2954 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction