Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSTZ1 Antibody (24E9.G9.G8), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP244177

Catalog No. NBP244177


Only null left
Add to Cart

Description

Description

Ensure accurate, reproducible results in Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page

GSTZ1 Monoclonal specifically detects GSTZ1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page.
Specifications

Specifications

GSTZ1
Monoclonal
1 mg/mL
Western Blot 1:500-1:1000, ELISA 1:50000-1:200000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, SDS-Page 1:100-1:2000
O43708
GSTZ1
This Protein A purified GSTZ1 Antibody (24E9.G9.G8) was prepared by repeated immunizations in mice with GSTGSTZ1 Protein (Uniprot: O43708)
Protein A purified
RUO
Primary
Reacts specifically with GSTZ1 protein.
Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, SDS-Page
24E9.G9.G8
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
EC 2.5.1.18, EC 5.2.1.2, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, Glutathione S-transferase zeta 1, glutathione transferase zeta 1, GSTZ1-1maleylacetoacetate isomerase, MAAIglutathione S-aryltransferase, MAI, maleylacetone isomerase, MGC2029, S-(hydroxyalkyl)glutathione lyase
Mouse
24.21 kDa
0.1 mg
metabolism
2954
Human, Mouse
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only