Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSTZ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100259
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100259 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100259 Supplier Invitrogen™ Supplier No. PA5100259
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GSTZ1.

This gene is a member of the glutathione S-transferase (GSTs) super-family which encodes multifunctional enzymes important in the detoxification of electrophilic molecules, including carcinogens, mutagens, and several therapeutic drugs, by conjugation with glutathione. This enzyme also plays a significant role in the catabolism of phenylalanine and tyrosine. Thus defects in this enzyme may lead to severe metabolic disorders including alkaptonuria, phenylketonuria and tyrosinaemia. Several transcript variants of this gene encode multiple protein isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GSTZ1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GSTZ1
Gene Accession No. O43708, P57113, Q9WVL0
Gene Alias 17-beta-HSD 10; 17-beta-hydroxysteroid dehydrogenase 10; 17b-HSD10; 2-methyl-3-hydroxybutyryl-CoA dehydrogenase; 3-hydroxy-2-methylbutyryl-CoA dehydrogenase; 3-hydroxyacyl-CoA dehydrogenase type II; 3-hydroxyacyl-CoA dehydrogenase type-2; ABAD; AB-binding alcohol dehydrogenase; amyloid-beta peptide binding alcohol dehydrogenase; CAMR; DUPXp11.22; Endoplasmic reticulum-associated amyloid beta-peptide-binding protein; ERAB; glutathione S-alkyltransferase; glutathione S-aralkyltransferase; glutathione S-aryltransferase; glutathione S-transferase alpha 3; glutathione S-transferase zeta 1; glutathione transferase zeta 1; glutathione transferase zeta 1 (maleylacetoacetate isomerase); GSTZ1; GSTZ1-1; HADH2; HCD2; HSD17B10; hydroxysteroid (17-beta) dehydrogenase 10; hydroxysteroid 17-beta dehydrogenase 10; hydroxysteroid dehydrogenase 10; MAAI; MAI; Maleylacetoacetate isomerase; maleylacetone isomerase; MHBD; Mitochondrial ribonuclease P protein 2; Mitochondrial RNase P protein 2; mitochondrial RNase P subunit 2; MRPP2; MRX17; MRX31; MRXS10; S-(hydroxyalkyl)glutathione lyase; SCHAD; SDR5C1; Short chain dehydrogenase/reductase family 5C member 1; short chain dehydrogenase/reductase family 5C, member 1; short chain L-3-hydroxyacyl-CoA dehydrogenase type 2; short chain type dehydrogenase/reductase XH98G2; Short-chain type dehydrogenase/reductase XH98G2; Type II HADH; XH98G2
Gene Symbols GSTZ1
Host Species Rabbit
Immunogen A synthesized peptide derived from human GSTZ1(Accession O43708), corresponding to amino acid residues P184-A216.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14874, 2954, 681913
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.