Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540504
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540504 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540504 Supplier Invitrogen™ Supplier No. PA540504
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: MSRSFYVDSL IIKDTSRPAP SLPEPHPGPD FFIPLGMPPP LVMSVSGPGC Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen GSX2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GSX2
Gene Accession No. Q9BZM3
Gene Alias genetic-screened homeobox 2; genomic screened homeo box 2; genomic screened homeo box 2 protein; GS homeobox 2; GSH2; Gsh-2; Gsx2; Homeobox protein GSH-2
Gene Symbols GSX2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GSH2 (aa 1-50).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170825
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.