Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GSX2 Recombinant Rabbit Monoclonal Antibody (HL2135)
GREENER_CHOICE

Catalog No. FEMA550394
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550394 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550394 Supplier Invitrogen™ Supplier No. MA550394
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

GSX2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen GSX2
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2135
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene GSX2
Gene Accession No. Q9BZM3
Gene Alias genetic-screened homeobox 2; genomic screened homeo box 2; genomic screened homeo box 2 protein; GS homeobox 2; GSH2; Gsh-2; Gsx2; Homeobox protein GSH-2
Gene Symbols GSX2
Host Species Rabbit
Immunogen Recombinant fragment of human GSX2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 170825, 364140
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.