Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GT 949
GREENER_CHOICESDP

Catalog No. 657850 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
65-785-0 50 mg
65-781-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 65-785-0 Supplier Tocris Bioscience™ Supplier No. 6578/50
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent and selective positive allosteric modulator of EAAT2

GT 949 is a potent and selective EAAT2 positive allosteric modulator (EC50 = 0.26 nM). Enhances the glutamate translocation rate, with no effect on substrate interaction. Enhances glutamate uptake by cultured astrocytes by approximately 58%. Exhibits no significant effect on DAT, SERT and NET or NMDA receptors. Neuroprotective in an in vitro model of excitotoxicity.

Specifications

Chemical Name or Material 3-((4-Cyclohexylpiperazin-1-yl)(1-phenethyl-1H-tetrazol-5-yl)methyl)-6-methoxyquinolin-2(1H)-one
CAS 460330-27-2
Quantity 50 mg
Molecular Formula C30H37N7O2
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.