Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTDC1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112953
Description
GTDC1 Polyclonal Antibody for Western Blot, ELISA

Specifications
GTDC1 | |
Polyclonal | |
Unconjugated | |
GTDC1 | |
E330008O22Rik; glycosyltransferase like domain containing 1; glycosyltransferase-like 1; glycosyltransferase-like domain containing 1; glycosyltransferase-like domain-containing protein 1; glycosyltransferase-like protein; Gtdc1; Hmat-Xa; mannosyltransferase candidate; mannosyltransferase-like protein Xa; mat-Xa | |
Rabbit | |
Protein G | |
RUO | |
79712 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
2 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q4AE62 | |
GTDC1 | |
Recombinant Human Glycosyltransferase-like domain-containing protein 1 protein (1-292AA). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction