Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2A1 Antibody (PCRP-GTF2A1-1F2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314054AF350
This item is not returnable.
View return policy
Description
GTF2A1 Monoclonal antibody specifically detects GTF2A1 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
GTF2A1 | |
Monoclonal | |
Alexa Fluor 350 | |
General transcription factor IIA subunit 1, general transcription factor IIA, 1, 19/37kDa, glucose regulated protein, 58kD pseudogene, MGC129969, MGC129970, TF2A1, TFIIA, TFIIA alpha, p55, TFIIA-42, TFIIAL, transcription initiation factor IIA subunit 1, Transcription initiation factor TFIIA 42 kDa subunit | |
Recombinant full-length human GTF2A1 protein (Uniprot: P52655) | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
2957 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-GTF2A1-1F2 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction