Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2A1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP146216PE
Description
GTF2A1 Polyclonal antibody specifically detects GTF2A1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot (Negative)Specifications
GTF2A1 | |
Polyclonal | |
Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Western Blot (Negative) | |
General transcription factor IIA subunit 1, general transcription factor IIA, 1, 19/37kDa, glucose regulated protein, 58kD pseudogene, MGC129969, MGC129970, TF2A1, TFIIA, TFIIA alpha, p55, TFIIA-42, TFIIAL, transcription initiation factor IIA subunit 1, Transcription initiation factor TFIIA 42 kDa subunit | |
The immunogen for this product maps to a region between residue 100 and 150 of human Transcription Initiation Factor IIA Subunit 1 using the numbering given in entry NP_056943.1 (GeneID 2957). | |
0.1 mL | |
DNA replication Transcription Translation and Splicing | |
2957 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry, Immunoprecipitation | |
PE | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction