Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530934 Supplier Invitrogen™ Supplier No. PA530934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, HeLa. Predicted reactivity: Mouse (96%), Rat (98%), Xenopus laevis (84%), Chicken (93%), Rhesus Monkey (100%), Bovine (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A) and several general initiation factors. One of these factors is TFIIA, which when purified from HeLa extracts consists of 35-, 19-, and 12-kD subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2A1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene GTF2A1
Gene Accession No. P52655
Gene Alias 6330549H03Rik; AA536742; AA959775; AW060250; general transcription factor 2a, 1; general transcription factor II A, 1; general transcription factor IIA 1; general transcription factor IIA subunit 1; general transcription factor IIA, 1; general transcription factor IIa, 1 (37kD and 19kD subunits); general transcription factor IIA, 1, 19/37kDa; glucose regulated protein, 58kD pseudogene; Gtf2a1; Tf2a; TF2A1; TFIIA; TFIIA alpha, p55, isoform 1; TFIIA alpha/beta; TFIIA alpha/beta subunits; TFIIA large subunit; TFIIA p19 subunit; TFIIA p35 subunit; Tfiia1; TFIIA-42; TfIIAa/b; TFIIAL; Transcription initiation factor IIA alpha chain; Transcription initiation factor IIA beta chain; Transcription initiation factor IIA subunit 1; transcription initiation factor TFIIA 42 kDa subunit
Gene Symbols GTF2A1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human GTF2A1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2957
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.