Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2A1L Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP169205
Description
GTF2A1L Polyclonal specifically detects GTF2A1L in Human samples. It is validated for Western Blot.Specifications
GTF2A1L | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
GTF2A1L | |
Synthetic peptides corresponding to GTF2A1L (general transcription factor IIA, 1-like) The peptide sequence was selected from the C terminal of GTF2A1L. Peptide sequence EFLGNIDGGDLKVPEEEADSISNEDSATNSSDNEDPQVNIVEEDPLNSGD. | |
Affinity purified | |
RUO | |
11036 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
1-like factor, general transcription factor IIA, 1-like, GTF2A1LF, MGC26254, TFIIA large subunit isoform ALF, TFIIA-alpha and beta-like factor, TFIIA-alpha/beta-like factor | |
Rabbit | |
49 kDa | |
100 μL | |
Primary | |
Equine: 86%; Mouse: 79%;. | |
Human, Mouse, Rat, Bovine, Canine, Equine | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction