Learn More
Invitrogen™ GTF2A1L Monoclonal Antibody (2E3)

Description
Peptide Sequence: PQVSQTNSNV ESVLSGSASM AQNLHDESLS TSPHGALHQH VTDIQLHILK NRMYGCDSVK QPRNIEEPSN IPVSEKDSNS QVDLSIRVTD DDIGEIIQ.
Specifications
Specifications
| Antigen | GTF2A1L |
| Applications | ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 2E3 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | GTF2A1L |
| Gene Accession No. | Q9UNN4 |
| Gene Alias | ALF; CXCDC1; general transcription factor II A, 1-like factor; general transcription factor IIA 1 like; general transcription factor IIA 1-like; general transcription factor IIA subunit 1 like; general transcription factor IIA, 1-like; GTF2A1L; GTF2A1LF; GTF2A1-like factor; KIAA2037; testis secretory sperm-binding protein Li 230m; TFIIA large subunit isoform ALF; TFIIA-alpha and beta-like factor; TFIIA-alpha/beta-like factor |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.