Learn More
Invitrogen™ GTF2A2 Polyclonal Antibody

Description
Antibody detects endogenous levels of total TF2A2.
Specifications
Specifications
| Antigen | GTF2A2 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | Gtf2a2 |
| Gene Accession No. | O08950, P52657, Q80ZM7 |
| Gene Alias | general transcription factor II A, 2; general transcription factor IIA 2; general transcription factor IIA subunit 2; general transcription factor IIA, 2; general transcription factor IIA, 2 (12kD subunit); general transcription factor IIA, 2, 12kDa; Gtf2a2; HsT18745; T18745; TF2A2; TFIIA; TFIIA gamma subunit; TFIIA p12 subunit; TFIIA-12; Tfiia2; TFIIA-gamma; TFIIAS; TfIIg; Transcription initiation factor IIA gamma chain; transcription initiation factor IIA subunit 2 |
| Gene Symbols | Gtf2a2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.