Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566005
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566005 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566005 Supplier Invitrogen™ Supplier No. PA566005
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AYQLYRNTTLG NSLQESLDEL IQSQQITPQL ALQVLLQFDK AINAALAQRV RNRVNFRGSL NTYRFCDNVW TFVLNDVEFR EVTELIKVDK VKIVACDGKN TGSNTTE Highest antigen sequence identity to the following orthologs - mouse 99%, rat 99%.

Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A; MIM 180660) and the general initiation factors TFIIA, TFIIB (MIM 189963), TFIID (MIM 313650), TFIIE (MIM 189962), TFIIF (MIM 189968), TFIIG/TFIIJ, and TFIIH (MIM 189972). The first step involves recognition of the TATA element by the TATA-binding subunit (TBP; MIM 600075) and may be regulated by TFIIA, a factor that interacts with both TBP and a TBP-associated factor (TAF; MIM 600475) in TFIID. TFIIA has 2 subunits (43 and 12 kD) in yeast and 3 subunits in higher eukaryotes. In HeLa extracts, it consists of a 35-kD alpha subunit and a 19-kD beta subunit encoded by the N- and C-terminal regions of GTF2A1 (MIM 600520), respectively, and a 12-kD gamma subunit encoded by GTF2A2 (DeJong et al., 1995).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2A2
Applications ChIP Assay, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gtf2a2
Gene Accession No. P52657
Gene Alias general transcription factor II A, 2; general transcription factor IIA 2; general transcription factor IIA subunit 2; general transcription factor IIA, 2; general transcription factor IIA, 2 (12kD subunit); general transcription factor IIA, 2, 12kDa; Gtf2a2; HsT18745; T18745; TF2A2; TFIIA; TFIIA gamma subunit; TFIIA p12 subunit; TFIIA-12; Tfiia2; TFIIA-gamma; TFIIAS; TfIIg; Transcription initiation factor IIA gamma chain; transcription initiation factor IIA subunit 2
Gene Symbols Gtf2a2
Host Species Rabbit
Immunogen Recombinant Human GTF2A2. Recombinant protein control fragment (Product #RP-105035).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2958
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.