Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501726389 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-6389 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-6389 Supplier Proteintech Group Inc Supplier No. 105401AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Accurate transcription initiation on TATA-containing class II genes involves the ordered assembly of RNA polymerase II (POLR2A), TFIID TFIIE TFIIF TFIIG/TFIIJ, and TFIIH respectively, and a 12-kD gamma subunit encoded by GTF2A2.

Specifications

Antigen GTF2A2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene GTF2A2
Gene Accession No. O08950, P52657, Q80ZM7
Gene Alias GTF2A2, HsT18745, TF2A2, TFIIA, TFIIA 12, TFIIA gamma, TFIIA p12 subunit, TFIIAS
Gene Symbols Gtf2a2
Host Species Rabbit
Immunogen GTF2A2 Fusion Protein Ag0825
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 235459, 2958, 83828
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.