Learn More
GTF2B Mouse anti-Human, Clone: OTI2A7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF808571

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes the general transcription factor IIB, one of the ubiquitous factors required for transcription initiation by RNA polymerase II. The protein localizes to the nucleus where it forms a complex (the DAB complex) with transcription factors IID and IIA. Transcription factor IIB serves as a bridge between IID, the factor which initially recognizes the promoter sequence, and RNA polymerase II.Specifications
GTF2B | |
Monoclonal | |
Unconjugated | |
GTF2B | |
TF2B, TFIIB | |
Mouse | |
Affinity Chromatography | |
RUO | |
2959 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2A7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q00403 | |
GTF2B | |
Human recombit protein fragment corresponding to amino acids 43-316 of human GTF2B produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.