Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2E1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566010 Supplier Invitrogen™ Supplier No. PA566010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QSKDHAATTAG AASLAGGHHR EAWATKGPSY EDLYTQNVVI NMDDQEDLHR ASLEGKSAKE RPIWLRESTV QGAYGSEDMK EGGIDMDAFQ EREEGHAGPD DNEEVMRALL IHEKKTSSAM AGSVGAAAPV TAANGSDSE Highest antigen sequence identity to the following orthologs - mouse 88%, rat 88%.

The general transcription factor TFIIE has an essential role in eukaryotic transcription initiation together with RNA polymerase II and other general factors. Human TFIIE consists of two subunits of relative molecular mass 57, 000 (TFIIE-alpha) and 34, 000 (TFIIE-beta) and joins the preinitiation complex after RNA polymerase II and TFIIF. TFIIE-alpha is necessary for transcription initiation together with TFIIE-beta, and recombinant TFIIE-alpha can fully replace the natural subunit in an in vitro transcription assay. Its sequence contains several interesting structural motifs (leucine repeat, zinc finger and helix-turn-helix) and sequence similarities to bacterial sigma factors that suggest direct involvement in the regulation of transcription initiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2E1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTF2E1
Gene Accession No. P29083
Gene Alias 2610024P03Rik; 56kDa; AI303800; FE; general transcription factor II E, polypeptide 1 (alpha subunit); general transcription factor II E, polypeptide 1 (alpha subunit, 56kDa); general transcription factor IIE 56 kDa subunit; General transcription factor IIE subunit 1; General transcription factor IIE subunit 1-like protein; general transcription factor IIE, polypeptide 1 (alpha subunit); general transcription factor IIE, polypeptide 1 (alpha subunit, 56kDa); general transcription factor IIE, polypeptide 1, alpha; general transcription factor IIE, polypeptide 1, alpha 56kDa; GTF2E1; TF2E1; TFIIE alpha subunit; TFIIE-A; TFIIE-alpha; Transcription initiation factor IIE subunit alpha
Gene Symbols GTF2E1
Host Species Rabbit
Immunogen Recombinant Human GTF2E1. Recombinant protein control fragment (Product #RP-105039).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2960
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.