Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2E2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597150
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597150 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597150 Supplier Invitrogen™ Supplier No. PA597150
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDPSLLRERE LFKKRALSTP VVEKRSASSE SSSSSSKKKK TKVEHGGSSG SKQNSDHSNG SFNLKALSGS SGYKFGVLAK IVNYMKTRHQ RGDTHPLTLD EILDETQHLD IGLKQKQWLM TEALVNNPKI EVIDGKYAFK PKYNVRDKKA LLRLLDQHDQ RGLGGILLED IEEALPNSQK AVKALGDQIL FVNRPDKKKI LFFNDKSCQF SVDEEFQKLW RSVTVDSMDE EKIEEYLKRQ GISSMQESGP KKVAPIQRRK KPASQKKRRF KTHNEHLAGV LKDYSDITSS K; Positive Samples: A-375, NIH/3T3, HT-1080, Jurkat, 293T, Mouse brain, Rat testis; Cellular Location: Nucleus.

The general transcription factor IIE (TFIIE) is part of the RNA polymerase II transcription initiation complex, recruiting TFIIH and being essential for promoter clearance by RNA polymerase II. TFIIE is a heterodimer (and sometimes heterotetramer) of alpha and beta subunits. The protein encoded by this gene represents the beta subunit of TFIIE.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2E2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.48 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GTF2E2
Gene Accession No. P29084, Q9D902
Gene Alias 34kDa; AI462509; C330006J08Rik; FE; general transcription factor II E, polypeptide 2 (beta subunit); general transcription factor II E, polypeptide 2 (beta subunit, 34kD); general transcription factor IIE subunit 2; general transcription factor IIE, polypeptide 2 (beta subunit); general transcription factor IIE, polypeptide 2 (beta subunit, 34kD); general transcription factor IIE, polypeptide 2, beta; general transcription factor IIE, polypeptide 2, beta 34kDa; Gtf2e2; T2EB; TF2E2; TFIIE beta subunit; TFIIE-B; TFIIE-beta; transcription initiation factor IIE subunit beta; TTD6
Gene Symbols GTF2E2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-291 of human GTF2E2 (NP_0020861).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2961, 306516, 68153
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.