Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2F1 Mouse anti-Human, Clone: OTI1C5, liquid, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene TA503373

Description
TFIIF is a general transcription initiation factor that binds to RNA polymerase II and helps to recruit it to the initiation complex in collaboration with TFIIB. It promotes transcription elongation.Specifications
GTF2F1 | |
Monoclonal | |
0.78 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P35269 | |
GTF2F1 | |
Full length human recombit protein of human GTF2F1 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI1C5 | |
Unconjugated | |
GTF2F1 | |
BTF4, RAP74, TF2F1, TFIIF | |
Mouse | |
Affinity Chromatography | |
RUO | |
2962 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction