Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2F1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554606 Supplier Invitrogen™ Supplier No. PA554606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LSNKKIYQEE EMPESGAGSE FNRKLREEAR RKKYGIVLKE FRPEDQPWLL RVNGKSGRKF KGIKKGGVTE NTSYYIFTQC PDGAFEAFPV HNWYNFTPLA RHRTLTAEEA EEEWERRNKV LNHFSIMQQR RLKDQDQD Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

TFIIF is a general transcription initiation factor that binds to RNA polymerase II and helps to recruit it to the initiation complex in collaboration with TFIIB. It promotes transcription elongation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2F1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gtf2f1
Gene Accession No. P35269
Gene Alias 2810405L04Rik; BTF4; C76800; General transcription factor IIF 74 kDa subunit; general transcription factor IIF polypeptide 1 (74kD subunit); general transcription factor IIF subunit 1; general transcription factor IIF, polypeptide 1; general transcription factor IIF, polypeptide 1, 74kDa; Gtf2f1; MGC94148; OTTHUMP00000237859; RAP74; TF2F1; TFIIF; TFIIF-alpha; transcription initiation factor IIF subunit alpha; Transcription initiation factor RAP74
Gene Symbols Gtf2f1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GTF2F1. Recombinant protein control fragment (Product #RP-102509).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2962
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.