Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2F2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540372
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540372 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540372 Supplier Invitrogen™ Supplier No. PA540372
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: KDLVDITKQP VVYLKEILKE IGVQNVKGIH KNTWELKPEY RHYQGEEKSD Sequence homology: Cow: 93%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 93%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.

GTF2F2 is required for both basal and HIV-1 Tat-activated transcription. GTF2F2 synergizes with HIV-1 Tat and the cellular co-activator Tat-SF1 during Tat-mediated transactivation of the HIV-1 LTR promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2F2
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GTF2F2
Gene Accession No. P13984
Gene Alias 1110031C13Rik; AI326315; ATP-dependent helicase GTF2F2; BTF4; General transcription factor IIF 30 kDa subunit; general transcription factor IIF subunit 2; general transcription factor IIF, polypeptide 2; general transcription factor IIF, polypeptide 2 (30kD subunit); general transcription factor IIF, polypeptide 2, 30kDa; GTF2F2; RAP30; TF2F2; TFIIF; TFIIF beta; TFIIF-beta; transcription factor gamma; transcription initiation factor IIF subunit beta; Transcription initiation factor RAP30; Unknown (protein for MGC:133695)
Gene Symbols GTF2F2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human GTF2F2 (aa 200-249).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2963
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.