Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2F2 Recombinant Rabbit Monoclonal Antibody (JE56-15)
GREENER_CHOICE

Catalog No. PIMA542489
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542489 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542489 Supplier Invitrogen™ Supplier No. MA542489
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Positive Control: Rat testis tissue lysate, A549 cell lysate, HepG2 cell lysate, HL-60 cell lysate, THP-1 cell lysate, Daudi cell lysate, rat testis tissue, human skin tissue, human placenta tissue, human uterus tissue. Subcellular Location: Nucleus.

GTF2F2 is required for both basal and HIV-1 Tat-activated transcription. GTF2F2 synergizes with HIV-1 Tat and the cellular co-activator Tat-SF1 during Tat-mediated transactivation of the HIV-1 LTR promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2F2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE56-15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 40% glycerol, 0.05% BSA and 0.05% sodium azide; pH 7.4
Gene GTF2F2
Gene Accession No. P13984, Q01750
Gene Alias 1110031C13Rik; AI326315; ATP-dependent helicase GTF2F2; BTF4; General transcription factor IIF 30 kDa subunit; general transcription factor IIF subunit 2; general transcription factor IIF, polypeptide 2; general transcription factor IIF, polypeptide 2 (30kD subunit); general transcription factor IIF, polypeptide 2, 30kDa; GTF2F2; RAP30; TF2F2; TFIIF; TFIIF beta; TFIIF-beta; transcription factor gamma; transcription initiation factor IIF subunit beta; Transcription initiation factor RAP30; Unknown (protein for MGC:133695)
Gene Symbols GTF2F2
Host Species Rabbit
Immunogen Synthetic peptide within C-terminal Human GTF2F2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2963, 81674
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.