Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2H1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | GTF2H1 |
---|---|
Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
GTF2H1 Polyclonal specifically detects GTF2H1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
GTF2H1 | |
Polyclonal | |
Rabbit | |
Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
Basic transcription factor 2 62 kDa subunit, BTF2 p62, BTF2general transcription factor IIH, polypeptide 1 (62kD subunit), General transcription factor IIH polypeptide 1, general transcription factor IIH subunit 1, general transcription factor IIH, polypeptide 1, 62kDa, TFB1, TFIIH, TFIIH basal transcription factor complex p62 subunit | |
GTF2H1 | |
IgG | |
Affinity Purified |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
P32780 | |
2965 | |
This antibody was developed against a recombinant protein corresponding to amino acids: PEGKAKIQLQLVLHAGDTTNFHFSNESTAVKERDAVKDLLQQLLPKFKRKANKELEEKNRMLQEDPVLFQLYKDLVVSQVISAEEFWANRLNVNATDSSSTSNHKQDVGISAAFLADVRPQTDGCNGLRYN | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title