Learn More
Invitrogen™ GTF2H1 Monoclonal Antibody (5B7)

Description
This antibody detects endogenous levels of GTF2H1 at a molecular weight of 62 kDa and does not cross-react with related proteins. Purity is >95% by SDS-PAGE.
Specifications
Specifications
| Antigen | GTF2H1 |
| Applications | Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | 5B7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | GTF2H1 |
| Gene Accession No. | P32780 |
| Gene Alias | 62kDa; AW743425; AW822074; Basic transcription factor 2 62 kDa subunit; BTF 2; BTF2; BTF2 p62; BTF2. TFIIH; BTF2-p62; C77871; general transcription factor II H, polypeptide 1; general transcription factor IIH polypeptide 1; General transcription factor IIH subunit 1; general transcription factor IIH, polypeptide 1; general transcription factor IIH, polypeptide 1 (62kD subunit); general transcription factor IIH, polypeptide 1, 62kDa; GTF2H1; P62; TF2H1; TFB1; TFIIH; TFIIH basal transcription factor complex p62 subunit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.