Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2H1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109797
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109797 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109797 Supplier Invitrogen™ Supplier No. PA5109797
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MATSSEEVLL IVKKVRQKKQ DGALYLMAER IAWAPEGKDR FTISHMYADI KCQKISPEGK AKIQLQLVLH AGDTTNFHFS NESTAVKERD AVKDLLQQLL PKFKRKANKE LEEKNRMLQE DPVLFQLYKD LVVSQVISAE EFWANRLNVN ATDSSSTSNH KQDVGISAAF LADVRPQTDG CNGLRYNLTS DIIESIFRTY PAVKMKYAEN VPHNMTEKEF WTRFFQSHYF HRDRLNTGSK DLFAECAKID EKGLKTMVSL GVKNPLLDLT ALEDKPLDEG YGISSVPSAS NSKSIKENSN AAIIKRFNHH SAMVLAAGLR KQEAQNEQTS EPSNMDGNSG DADCFQPAVK RAKLQESIEY EDLGKNNSVK TIALNLKKSD RYYHGPTPIQ SLQYATSQDI INSFQSIRQE MEAYTPKLTQ VLSSS.

GTF2H1 (general transcription factor IIH, polypeptide 1, 62kDa) is a component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA. When complexed to CAK, GTF2H1 is involved in RNA transcription by RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2H1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.87 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GTF2H1
Gene Accession No. P32780, Q9DBA9
Gene Alias 62kDa; AW743425; AW822074; Basic transcription factor 2 62 kDa subunit; BTF 2; BTF2; BTF2 p62; BTF2. TFIIH; BTF2-p62; C77871; general transcription factor II H, polypeptide 1; general transcription factor IIH polypeptide 1; General transcription factor IIH subunit 1; general transcription factor IIH, polypeptide 1; general transcription factor IIH, polypeptide 1 (62kD subunit); general transcription factor IIH, polypeptide 1, 62kDa; GTF2H1; P62; TF2H1; TFB1; TFIIH; TFIIH basal transcription factor complex p62 subunit
Gene Symbols GTF2H1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-425 of human GTF2H1 (NP_001135779.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14884, 2965, 361580
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.