Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2H1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA536246
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA536246 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA536246 Supplier Invitrogen™ Supplier No. PA536246
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody detects endogenous protein at a molecular weight of 62 kDa. Purity is >95% by SDS-PAGE.

GTF2H1 (general transcription factor IIH, polypeptide 1, 62kDa) is a component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA. When complexed to CAK, GTF2H1 is involved in RNA transcription by RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2H1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide; pH 7.2
Gene GTF2H1
Gene Accession No. P32780, Q9DBA9
Gene Alias 62kDa; AW743425; AW822074; Basic transcription factor 2 62 kDa subunit; BTF 2; BTF2; BTF2 p62; BTF2. TFIIH; BTF2-p62; C77871; general transcription factor II H, polypeptide 1; general transcription factor IIH polypeptide 1; General transcription factor IIH subunit 1; general transcription factor IIH, polypeptide 1; general transcription factor IIH, polypeptide 1 (62kD subunit); general transcription factor IIH, polypeptide 1, 62kDa; GTF2H1; P62; TF2H1; TFB1; TFIIH; TFIIH basal transcription factor complex p62 subunit
Gene Symbols GTF2H1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids N-terminus of human TFIIH p62.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14884, 2965, 361580
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.