Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2H1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP304722100UL
Description
GTF2H1 Polyclonal specifically detects GTF2H1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
GTF2H1 | |
Polyclonal | |
PBS with 50% glycerol, pH7.3. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
Unconjugated | |
Basic transcription factor 2 62 kDa subunit, BTF2 p62, BTF2general transcription factor IIH, polypeptide 1 (62kD subunit), General transcription factor IIH polypeptide 1, general transcription factor IIH subunit 1, general transcription factor IIH, polypeptide 1, 62kDa, TFB1, TFIIH, TFIIH basal transcription factor complex p62 subunit | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-425 of human GTF2H1 (NP_001135779.1). | |
100 μL | |
Cell Cycle and Replication, DNA Repair, Nucleotide Excision Repair | |
2965 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction