Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2H2 Antibody (PCRP-GTF2H2-1B9), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308257AF488
Description
GTF2H2 Monoclonal antibody specifically detects GTF2H2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein ArraySpecifications
GTF2H2 | |
Monoclonal | |
Alexa Fluor 488 | |
Basic transcription factor 2 44 kDa subunit, BTF2, BTF2 p44, BTF2P44TFIIH basal transcription factor complex p44 subunit, General transcription factor IIH polypeptide 2, general transcription factor IIH subunit 2, general transcription factor IIH, polypeptide 2 (44kD subunit), general transcription factor IIH, polypeptide 2, 44kD subunit, general transcription factor IIH, polypeptide 2, 44kDa, MGC102806, T-BTF2P44, TFIIH | |
Recombinant full-length human GTF2H2 protein (Uniprot: Q13888) | |
100 μg | |
Stem Cell Markers | |
2966 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, Immunofluorescence, Protein Array | |
PCRP-GTF2H2-1B9 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction