Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2H2 Polyclonal Antibody
GTF2H2 Polyclonal Antibody
Supplier: Thermo Scientific PA536267
Description
GTF2H2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
GTF2H2 | |
Polyclonal | |
Unconjugated | |
Gtf2h2 | |
44kDa; basal transcription factor 2, p44 subunit; basic transcription factor 2 44 kDa subunit; BTF2; BTF2 p44; BTF2P44; BTF2-p44; general transcription factor II H, polypeptide 2; general transcription factor II H, polypeptide 2 (44 kDa subunit); general transcription factor IIH polypeptide 2; General transcription factor IIH subunit 2; general transcription factor IIH, polypeptide 2; general transcription factor IIH, polypeptide 2 (44 kDa subunit); general transcription factor IIH, polypeptide 2, 44kD subunit; general transcription factor IIH, polypeptide 2, 44kDa; Gtf2h2; p44; T-BTF2P44; TFIIH; TFIIH basal transcription factor complex p44 subunit | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23894, 294693, 2966 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
A0JN27, Q13888, Q9JIB4 | |
Gtf2h2 | |
Synthetic peptide corresponding to the N-terminal of human TFIIH p44. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction