Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2H2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565462
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565462 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565462 Supplier Invitrogen™ Supplier No. PA565462
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HVSPPPASSSS ECSLIRMGFP QHTIASLSDQ DAKPSFSMAH LDGNTEPGLT LGGYFCPQCR AKYCELPVEC KICG Highest antigen sequence identity to the following orthologs - mouse 97%, rat 96%.

Initiation of transcription from protein-coding genes in eukaryotes is a complex process that requires RNA polymerase II, as well as families of basal transcription factors. Binding of the factor TFIID (TBP) to the TATA box is believed to be the first step in the formation of a multiprotein complex containing several additional factors, including TFIIA, TFIIB, TFIIE, TFIIF and TFII. TFIIH (or BTF2) is a multisubunit transcription/DNA repair factor that possesses several enzymatic activities. The core of TFIIH is composed of five subunits, designated p89 (XPB or ERCC3), p62, p52, p44 and p34. Additional subunits of the TFIIH complex are p80 (XPD or ERCC2) and the ternary kinase complex composed of Cdk7, cyclin H and MAT1. Both p89 and p80 have ATP-dependent helicase activity. The p62, p52 and p44 subunits have been shown to be involved in nucleotide excision repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2H2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Gtf2h2
Gene Accession No. Q13888
Gene Alias 44kDa; basal transcription factor 2, p44 subunit; basic transcription factor 2 44 kDa subunit; BTF2; BTF2 p44; BTF2P44; BTF2-p44; general transcription factor II H, polypeptide 2; general transcription factor II H, polypeptide 2 (44 kDa subunit); general transcription factor IIH polypeptide 2; General transcription factor IIH subunit 2; general transcription factor IIH, polypeptide 2; general transcription factor IIH, polypeptide 2 (44 kDa subunit); general transcription factor IIH, polypeptide 2, 44kD subunit; general transcription factor IIH, polypeptide 2, 44kDa; Gtf2h2; p44; T-BTF2P44; TFIIH; TFIIH basal transcription factor complex p44 subunit
Gene Symbols Gtf2h2
Host Species Rabbit
Immunogen Recombinant Human GTF2H2. Recombinant protein control fragment (Product #RP-104829).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2966
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.