Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTF2H2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5104234
Description
Antibody detects endogenous levels of total TF2H2.
Initiation of transcription from protein-coding genes in eukaryotes is a complex process that requires RNA polymerase II, as well as families of basal transcription factors. Binding of the factor TFIID (TBP) to the TATA box is believed to be the first step in the formation of a multiprotein complex containing several additional factors, including TFIIA, TFIIB, TFIIE, TFIIF and TFII. TFIIH (or BTF2) is a multisubunit transcription/DNA repair factor that possesses several enzymatic activities. The core of TFIIH is composed of five subunits, designated p89 (XPB or ERCC3), p62, p52, p44 and p34. Additional subunits of the TFIIH complex are p80 (XPD or ERCC2) and the ternary kinase complex composed of Cdk7, cyclin H and MAT1. Both p89 and p80 have ATP-dependent helicase activity. The p62, p52 and p44 subunits have been shown to be involved in nucleotide excision repair.
Specifications
GTF2H2 | |
Polyclonal | |
Unconjugated | |
Gtf2h2 | |
44kDa; basal transcription factor 2, p44 subunit; basic transcription factor 2 44 kDa subunit; BTF2; BTF2 p44; BTF2P44; BTF2-p44; general transcription factor II H, polypeptide 2; general transcription factor II H, polypeptide 2 (44 kDa subunit); general transcription factor IIH polypeptide 2; General transcription factor IIH subunit 2; general transcription factor IIH, polypeptide 2; general transcription factor IIH, polypeptide 2 (44 kDa subunit); general transcription factor IIH, polypeptide 2, 44kD subunit; general transcription factor IIH, polypeptide 2, 44kDa; Gtf2h2; p44; T-BTF2P44; TFIIH; TFIIH basal transcription factor complex p44 subunit | |
Rabbit | |
Affinity chromatography | |
RUO | |
23894, 294693, 2966 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
A0JN27, Q13888, Q9JIB4 | |
Gtf2h2 | |
A synthesized peptide derived from human GTF2H2(Accession Q13888), corresponding to amino acid residues M1-E48. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction