Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2H2C Antibody (PCRP-GTF2H2C-2C9), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308896AF532
Description
GTF2H2C Monoclonal antibody specifically detects GTF2H2C in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein ArraySpecifications
GTF2H2C | |
Monoclonal | |
Alexa Fluor 532 | |
General transcription factor IIH polypeptide 2-like protein, general transcription factor IIH subunit 2-like protein, general transcription factor IIH, polypeptide 2C, GTF2H2C_2 | |
Recombinant full-length human GTF2H2C protein (Uniprot: Q6P1K8) | |
100 μg | |
Direct Reversal of DNA Damage | |
728340 | |
Store at 4°C in the dark. | |
IgG2c |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PCRP-GTF2H2C-2C9 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction