Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2H2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548199
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548199 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548199 Supplier Invitrogen™ Supplier No. PA548199
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GTF2H2C Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2H2C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GTF2H2C
Gene Accession No. Q6P1K8
Gene Alias general transcription factor IIH polypeptide 2-like protein; General transcription factor IIH subunit 2-like protein; general transcription factor IIH, polypeptide 2C; GTF2H2 family member C; GTF2H2C; GTF2H2C_2; GTF2H2D
Gene Symbols GTF2H2C
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 314-343 amino acids from the C-terminal region of human GTF2H2C.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728340
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.