Learn More
Invitrogen™ GTF2H2C Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA592359
Description
Immunogen sequence: MDEEPERTKR WEGGYERTWE ILKEDESGSL KATIEDILFK AKRKRVFEHH GQVRLGMMRH LYVVVDGSRT MEDQDLKPNR LTCTLKLLEY FVEEYFDQNP ISQIGIIVTK SKRAEKLTEL SGNPRKHITS LKEAVDMTCH GEPSLYNSLS MAMQTLKHMP GHTSREVLII FSSLTTCDPS NIYDLIKTLK AAKIRVSVIG LSAEVRVCTV LARETGGTYH VILDESHYKE LLTHHLSPPP ASSSSECSLI RMGFPQHTIA SLSDQDAKPS FSMAHLDGNT EPGLTLGGYF CPQCRAKYCE; Positive Samples: LO2; Cellular Location: Nucleus.
Component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
Specifications
GTF2H2C | |
Polyclonal | |
Unconjugated | |
GTF2H2C | |
general transcription factor IIH polypeptide 2-like protein; General transcription factor IIH subunit 2-like protein; general transcription factor IIH, polypeptide 2C; GTF2H2 family member C; GTF2H2C; GTF2H2C_2; GTF2H2D | |
Rabbit | |
Affinity chromatography | |
RUO | |
728340 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.46 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P1K8 | |
GTF2H2C | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GTF2H2C (NP_0010921981). | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.