Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2H2C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109798
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109798 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109798 Supplier Invitrogen™ Supplier No. PA5109798
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDEEPERTKR WEGGYERTWE ILKEDESGSL KATIEDILFK AKRKRVFEHH GQVRLGMMRH LYVVVDGSRT MEDQDLKPNR LTCTLKLLEY FVEEYFDQNP ISQIGIIVTK SKRAEKLTEL SGNPRKHITS LKEAVDMTCH GEPSLYNSLS MAMQTLKHMP GHTSREVLII FSSLTTCDPS NIYDLIKTLK AAKIRVSVIG LSAEVRVCTV LARETGGTYH VILDESHYKE LLTHHLSPPP ASSSSECSLI RMGFPQHTIA.

Component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2H2C
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 4.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GTF2H2C
Gene Accession No. Q6P1K8
Gene Alias general transcription factor IIH polypeptide 2-like protein; General transcription factor IIH subunit 2-like protein; general transcription factor IIH, polypeptide 2C; GTF2H2 family member C; GTF2H2C; GTF2H2C_2; GTF2H2D
Gene Symbols GTF2H2C
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human GTF2H2C_2 (NP_001035955.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 728340
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.