Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTF2H3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540374
Description
Peptide sequence: VIASHIQESR FLYPGKNGRL GDFFGDPGNP PEFNPSGSKD GKYELLTSAN Sequence homology: Cow: 92%; Guinea Pig: 92%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
GTF2H3 interacts with HIV-1 Tat as a component of the HIV-1 transcription pre-initiation complex, but is released from the elongation complex which includes P-TEFb. It synergizes with HIV-1 Tat to induce transcription elongation from the HIV-1 LTR promoter.
Specifications
GTF2H3 | |
Polyclonal | |
Unconjugated | |
GTF2H3 | |
34kDa; 5033417D07Rik; Basic transcription factor 2 34 kDa subunit; BTF2; BTF2 p34; BTF2-p34; C730029A10; D5Ertd679e; General transcription factor IIH polypeptide 3; general transcription factor IIH subunit 3; general transcription factor IIH, polypeptide 3; general transcription factor IIH, polypeptide 3, 34kDa; Gtf2h3; P34; TFB4; TFIIH; TFIIH basal transcription factor complex p34 subunit | |
Rabbit | |
Affinity Chromatography | |
RUO | |
2967 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q13889 | |
GTF2H3 | |
Synthetic peptide directed towards the N-terminal of human GTF2H3 (aa 59-108). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction