Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2H3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577161
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577161 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577161 Supplier Invitrogen™ Supplier No. PA577161
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

GTF2H3 interacts with HIV-1 Tat as a component of the HIV-1 transcription pre-initiation complex, but is released from the elongation complex which includes P-TEFb. It synergizes with HIV-1 Tat to induce transcription elongation from the HIV-1 LTR promoter.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2H3
Applications Immunoprecipitation, Western Blot, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene GTF2H3
Gene Accession No. Q13889, Q8VD76
Gene Alias 34kDa; 5033417D07Rik; Basic transcription factor 2 34 kDa subunit; BTF2; BTF2 p34; BTF2-p34; C730029A10; D5Ertd679e; General transcription factor IIH polypeptide 3; general transcription factor IIH subunit 3; general transcription factor IIH, polypeptide 3; general transcription factor IIH, polypeptide 3, 34kDa; Gtf2h3; P34; TFB4; TFIIH; TFIIH basal transcription factor complex p34 subunit
Gene Symbols GTF2H3
Host Species Rabbit
Immunogen Recombinant full length Human GTF2H3.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 209357, 2967
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.