Learn More
Invitrogen™ GTF2H3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596603
Description
Immunogen sequence: MVSDEDELNL LVIVVDANPI WWGKQALKES QFTLSKCIDA VMVLGNSHLF MNRSNKLAVI ASHIQESRFL YPGKNGRLGD FFGDPGNPPE FNPSGSKDGK YELLTSANEV IVEEIKDLMT KSDIKGQHTE TLLAGSLAKA LCYIHRMNKE VKDNQEMKSR ILVIKAAEDS ALQYMNFMNV IFAAQKQNIL IDACVLDSDS GLLQQACDIT GGLYLKVPQM PSLLQYLLWV FLPDQDQRSQ LILPPPVHVD YRAACFCHRN LIEIGYVCSV CLSIFCNFSP ICTTCETAFK ISLPPVLKAK KKKLKVSA; Positive Samples: HL-60, HT-29, MCF-7, Jurkat; Cellular Location: Nucleus.
GTF2H3 interacts with HIV-1 Tat as a component of the HIV-1 transcription pre-initiation complex, but is released from the elongation complex which includes P-TEFb. It synergizes with HIV-1 Tat to induce transcription elongation from the HIV-1 LTR promoter.
Specifications
GTF2H3 | |
Polyclonal | |
Unconjugated | |
GTF2H3 | |
34kDa; 5033417D07Rik; Basic transcription factor 2 34 kDa subunit; BTF2; BTF2 p34; BTF2-p34; C730029A10; D5Ertd679e; General transcription factor IIH polypeptide 3; general transcription factor IIH subunit 3; general transcription factor IIH, polypeptide 3; general transcription factor IIH, polypeptide 3, 34kDa; Gtf2h3; P34; TFB4; TFIIH; TFIIH basal transcription factor complex p34 subunit | |
Rabbit | |
Affinity Chromatography | |
RUO | |
209357, 2967 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunoprecipitation, Western Blot | |
2.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q13889, Q8VD76 | |
GTF2H3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-308 of human GTF2H3 (NP_0015072). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.