Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2H4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA597048
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597048 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597048 Supplier Invitrogen™ Supplier No. PA597048
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MESTPSRGLN RVHLQCRNLQ EFLGGLSPGV LDRLYGHPAT CLAVFRELPS LAKNWVMRML FLEQPLPQAA VALWVKKEFS KAQEESTGLL SGLRIWHTQL LPGGLQGLIL NPIFRQNLRI ALLGGGKAWS DDTSQLGPDK HARDVPSLDK YAEERWEVVL HFMVGSPSAA VSQDLAQLLS QAGLMKSTEP GEPPCITSAG FQFLLLDTPA QLWYFMLQYL QTAQSRGMDL VEILSFLFQL SFSTLGKDYS VEGMSDSLLN FLQHLREFGL VFQRKRKSRR YYPTRLAINL SSGVSGAGGT; Positive Samples: HL-60, HeLa, Mouse brain, Mouse skeletal muscle; Cellular Location: Nucleus.

GTF2H4 belongs to the TFB2 family. It is a component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2H4
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GTF2H4
Gene Accession No. O70422, Q92759
Gene Alias AW545633; basic transcription factor 2 52 kDa subunit; BTF2 p52; general transcription factor II H, polypeptide 4; general transcription factor IIH polypeptide 4; General transcription factor IIH subunit 4; general transcription factor IIH, polypeptide 4, 52kDa; GTF2H4; p52; TFB2; TFIIH; TFIIH basal transcription factor complex p52 subunit; TFIIH transcription/DNA repair factor p52 subunit
Gene Symbols GTF2H4
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GTF2H4 (NP_0015081).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14885, 2968
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.