Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GTF2H4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA540568
Description
Peptide sequence: ESTPSRGLNR VHLQCRNLQE FLGGLSPGVL DRLYGHPATC LAVFRELPSL Sequence homology: Cow: 92%; Dog: 100%; Goat: 92%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
GTF2H4 belongs to the TFB2 family. It is a component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
Specifications
GTF2H4 | |
Polyclonal | |
Unconjugated | |
GTF2H4 | |
AW545633; basic transcription factor 2 52 kDa subunit; BTF2 p52; general transcription factor II H, polypeptide 4; general transcription factor IIH polypeptide 4; General transcription factor IIH subunit 4; general transcription factor IIH, polypeptide 4, 52kDa; GTF2H4; p52; TFB2; TFIIH; TFIIH basal transcription factor complex p52 subunit; TFIIH transcription/DNA repair factor p52 subunit | |
Rabbit | |
Affinity chromatography | |
RUO | |
2968 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q92759 | |
GTF2H4 | |
Synthetic peptide directed towards the N-terminal of human GTF2H4 (aa 2-51). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction