Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2H4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103346
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103346 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103346 Supplier Invitrogen™ Supplier No. PA5103346
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GTF2H4.

GTF2H4 belongs to the TFB2 family. It is a component of the core-TFIIH basal transcription factor involved in nucleotide excision repair (NER) of DNA and, when complexed to CAK, in RNA transcription by RNA polymerase II.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2H4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GTF2H4
Gene Accession No. O70422, Q92759
Gene Alias AW545633; basic transcription factor 2 52 kDa subunit; BTF2 p52; general transcription factor II H, polypeptide 4; general transcription factor IIH polypeptide 4; General transcription factor IIH subunit 4; general transcription factor IIH, polypeptide 4, 52kDa; GTF2H4; p52; TFB2; TFIIH; TFIIH basal transcription factor complex p52 subunit; TFIIH transcription/DNA repair factor p52 subunit
Gene Symbols GTF2H4
Host Species Rabbit
Immunogen A synthesized peptide derived from human GTF2H4(Accession Q92759), corresponding to amino acid residues L108-V158.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14885, 294236, 2968
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.