Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2H4 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$181.90 - $487.50
Specifications
Antigen | GTF2H4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
GTF2H4 Polyclonal specifically detects GTF2H4 in Human, Mouse samples. It is validated for Western Blot.Specifications
GTF2H4 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse | |
Basic transcription factor 2 52 kDa subunit, BTF2 p52, General transcription factor IIH polypeptide 4, general transcription factor IIH subunit 4, general transcription factor IIH, polypeptide 4 (52kD subunit), general transcription factor IIH, polypeptide 4, 52kDa, TFB2, TFIIH, TFIIH basal transcription factor complex p52 subunit | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human GTF2H4 (NP_001508.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
Rabbit | |
DNA Repair, Nucleotide Excision Repair | |
PBS with 50% glycerol, pH7.3. | |
2968 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title