Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GTF2I Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA582928
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA582928 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA582928 Supplier Invitrogen™ Supplier No. PA582928
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: FVIKKHELLN STREDLQLDK PASGVKEEWY ARITKLRKMV DQLFCKKFAE ALGSTEAKAV PYQKFEAHPN DLYVEGLPEN IPFRSPSWYG IPRLEKIIQV GNRIKFVIKR PELLTHSTTE VTQPRTNTPV KEDWNVRIT Highest antigen sequence indentity to the following orthologs: Rat - 99%, Mouse - 100%.

This gene encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11. 23. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7, 13 and 21.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2I
Applications ChIP Assay, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GTF2I
Gene Accession No. P78347
Gene Alias 6030441I21Rik; BAP135; BAP-135; Bruton tyrosine kinase-associated protein 135; BTKAP1; BTK-associated protein 135; BTK-associated protein, 135kD; DIWS; Diws1t; general transcription factor II I; general transcription factor IIi; general transcription factor II-I; Gtf2i; Gtf2ird1; GTFII-I; IB291; OTTHUMP00000209509; Spin; SRF-Phox1-interacting protein; tfiii; TFII-I; WBS; WBSCR6; Williams-Beuren syndrome chromosomal region 6 protein; Williams-Beuren syndrome chromosome region 6
Gene Symbols GTF2I
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GTF2I. Recombinant protein control fragment (Product #RP-102472).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2969
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.