Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2IRD1 Antibody, DyLight 550, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10061055R
Description
GTF2IRD1 Polyclonal antibody specifically detects GTF2IRD1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
GTF2IRD1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
BEN, CREAM1, General transcription factor III, general transcription factor II-I repeat domain-containing protein 1, GTF2I repeat domain containing 1, GTF2I repeat domain-containing protein 1, GTF3GTF2I repeat domain-containing 1, hMusTRD1alpha1, Muscle TFII-I repeat domain-containing protein 1, muscle TFII-I repeat domain-containing protein 1 alpha 1, MusTRD1, MusTRD1/BEN, MUSTRD1general transcription factor 3, RBAP2WBSCR12binding factor for early enhancer, Slow-muscle-fiber enhancer-binding protein, USE B1-binding protein, WBS, WBSCR11, Williams-Beuren syndrome chromosomal region 11 protein, Williams-Beuren syndrome chromosomal region 12 protein, Williams-Beuren syndrome chromosome region 11 | |
The immunogen recognized by this antibody maps to a region between residue 650 and 700 of human general transcription factor 2I repeat domain containing 1 using the numbering given in entry NP_057412.1 (GeneID 9569). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
DyLight 550 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
9569 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction