Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GTF2IRD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA541578
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA541578 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA541578 Supplier Thermo Scientific Supplier No. PA541578
Add to Cart
Add to Cart

GTF2IRD1 Polyclonal Antibody

GTF2IRD1 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen GTF2IRD1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GTF2IRD1
Gene Accession No. Q9UHL9
Gene Alias 1700012P16Rik; Alb/c-myc line 166.8; Alb-c-myc line 166.8; BEN; Binding factor for early enhancer; c-myc line 166.8; CREAM1; ESTM9; general transcription factor 3; general transcription factor II I repeat domain-containing 1; General transcription factor III; general transcription factor II-I repeat domain-containing protein 1; GTF2I repeat domain containing 1; GTF2I repeat domain-containing 1; GTF2I repeat domain-containing protein 1; Gtf2il; Gtf2ird1; GTF3; hMusTRD1alpha1; Muscle TFII-I repeat domain-containing protein 1; muscle TFII-I repeat domain-containing protein 1 alpha 1; MUSTRD1; MusTRD1/BEN; RBAP2; slow-muscle-fiber enhancer-binding protein; Tg(Alb1-Myc)166.8Sst; transcription factor GTF3 alpha 2; transcription factor GTF3 gamma 2; USE B1-binding protein; WBS; WBSCR11; WBSCR12; Williams-Beuren syndrome chromosomal region 11 protein; Williams-Beuren syndrome chromosomal region 12 protein; Williams-Beuren syndrome chromosome region 11; X83320
Gene Symbols GTF2IRD1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GTF2IRD1 (aa 1-50).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9569
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.