Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GTF2IRD2 Antibody (PCRP-GTF2IRD2-1B12), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308512MFV500
Description
GTF2IRD2 Monoclonal antibody specifically detects GTF2IRD2 in Human samples. It is validated for Flow CytometrySpecifications
GTF2IRD2 | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
84163 | |
Store at 4C in the dark. | |
IgG2b |
Flow Cytometry | |
PCRP-GTF2IRD2-1B12 | |
Flow Cytometry | |
FLJ21423, FP630, general transcription factor II i repeat domain 2 alpha, general transcription factor II-I repeat domain-containing protein 2A, GTF2I repeat domain containing 2, GTF2I repeat domain-containing protein 2A, GTF2IRD2 alpha, GTF2IRD2AFLJ37938, MGC75203, transcription factor GTF2IRD2, Transcription factor GTF2IRD2-alpha | |
Recombinant full-length human GTF2IRD2 protein (Uniprot: Q86UP8) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction